D(+)-sakkaroosi, kiteinen, food grade

Toimittaja: MP Biomedicals

0290471305 0290471325 0290471380
ICNA0290471305EA 51 EUR
ICNA0290471305 ICNA0290471325 ICNA0290471380
D(+)-sakkaroosi, kiteinen, food grade
D(+)-sakkaroosi

Soluble in water (1 g/0,5 ml at 25 °C; 1 g/0,2 ml boiling water), ethanol (1 g/170 ml) or methanol (1 g/100 ml); moderately soluble in glycerol or pyridine.

Kaava: C₁₂H₂₂O₁₁
Sulamispiste: 177…187 °C
MDL numero: MFCD00006626
CAS-numero: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR