Polyetyleeniglykoli 8000, jauhe

Toimittaja: MP Biomedicals

02195445.5 0219544501 0219544502
ICNA02195445.5EA 45.2 EUR
ICNA02195445.5 ICNA0219544501 ICNA0219544502
Polyetyleeniglykoli 8000, jauhe
Polyetyleeniglykoli 8000

Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water.


PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridisation. PEG 8 000, can be used to precipitate DNA. PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis.


PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds. PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives


Soluble in aqueous solution (50 mg/ml - clear, colorless solution).

Kaava: H(OCH₂CH₂)nOH
MDL numero: MFCD01779615
CAS-numero: 25322-68-3

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
Melting Point 56 - 66 °C
pH (5% aq soln) 5.0 - 7.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR