L(-)-tyrosiini, off-white powder cell culture reagent

Toimittaja: MP Biomedicals

0219475950 0219475980 0219475990
ICNA0219475950EA 96.4 EUR
ICNA0219475950 ICNA0219475980 ICNA0219475990
L(-)-tyrosiini, off-white powder cell culture reagent
L(-)-tyrosiini

Storage: Store at room temperature (15-30 °C)
L-Tyrosine is one of the three aromatic amino acids, and is formed from the hydroxylation of phenylalanine.
L-Tyrosine is used in cell culture media and is a component of MEM amino acids solution. L-Tyrosine has been used in a cell culture study of the amino acid transport system b0,+ in epithelial cells isolated from chicken jejunum.


  • Cell Culture Reagent Grade
  • Extinction Coefficient (E1%): EmM = 8.2 (223 nm), 1.4 (274.5 nm), in 0.1 M HCl; 11.05 (240 nm), 2.33 (293 nm), in 0.1 N NaOH (Lit.)
  • Isoelectric Point (pl): 5.63 (Lit.)
  • pKa: 2.20 (-COOH), 9.21 (a-NH2), 10.46 (phenolic OH) (Lit.)
  • Solubility in water ~45mg/100 mL. Soluble in alkaline and acidic aqueous solution.

Kaava: C₉H₁₁NO₃
Sulamispiste: 342…344 °C (kirj.)
Tiheys: 1,4 g/mL (kirj.)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002606
CAS-numero: 60-18-4

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
UV/VIS: I max (water) 270 - 275 nm
Loss on Drying <0.5%
Optical Rotation (c=4, 1N HCl) -9.0° to -12.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR