Tiamiinihydrokloridi, powder

Toimittaja: MP Biomedicals

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

0219474983 0219474925 0219474980
ICNA0219474983EA 246 EUR
ICNA0219474983 ICNA0219474925 ICNA0219474980
Tiamiinihydrokloridi, powder
Tiamiinihydrokloridi

Used in fluorometric determination of mercury. Required to support energy metabolism and amino acid synthesis in cultured cells. Present in many classical and serum-free formulations.


Varoitus: Research use only (RUO). Ready for CE IVD labeling of clinical applications.

Kaava: C₁₂H₁₈Cl₂N₄OS
Sulamispiste: 164 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012780
CAS-numero: 67-03-8
EINECS: 200-641-8

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
pH 2,7 - 3,4 (1% aq soln)
UV/VIS: OD400 ≤0,25 (1% aq soln)
Loss on Drying ≤5,0%
Residue on Ignition ≤0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR