Nitro blue tetrazolium molekyylibiologiaan

Toimittaja: MP Biomedicals
Varoitus

0219399950 0219399991 0219399983
ICNA0219399950EA 30.6 EUR
ICNA0219399950 ICNA0219399991 ICNA0219399983
Nitro blue tetrazolium molekyylibiologiaan
Nitro blue tetrazolium

p-Nitroblue Tetrazolium Chloride is an NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase) (IC50 = 3-4 μM). In combination with 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and an appropriate reaction buffer, Nitroblue tetrazolium forms a substrate for the colorimetric detection of alkaline phosphatase conjugates in a variety of molecular and cell biology techniques.

p-nitro blue tetrazolium is used for the detection of alkaline phosphatase on western blots. It has also been used as a redox indicator for other enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels, and pentose shunt dehydrogenases. Redox and halfwave potentials have been determined for p - nitro blue tetrazolium. It has also been used as a colorimetric indicator of bacterial infection in blood samples.


  • Solubility: Slightly (10 mg/mL) soluble in water.


Storage temperature: + 4 °C

Kaava: C₄₀H₃₀Cl₂N₁₀O₆
Sulamispiste: 189…192 °C (haj. lämpötila, lit.)
Storage Temperature: Jääkaappi
MDL numero: MFCD00012159
CAS-numero: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR