Bentsyyli-alkoholi, clear liquid ACS

Toimittaja: MP Biomedicals
Varoitus

0219388290 0219388280
ICNA0219388290EA 115 EUR
ICNA0219388290 ICNA0219388280
Bentsyyli-alkoholi, clear liquid ACS
Bentsyyli-alkoholi

Storage: Store at Room Temperature (15-30 °C)
Benzyl alcohol (BnOH) is a monoaromatic primary alcohol widely used as a solvent and an intermediate in cosmetic formulations, pharmaceutical and flavor/fragrance industries. It shows bacteriostatic property. Commercial method for its synthesis is by the alkaline hydrolysis of benzyl chloride. Conformational studies by ab initio calculations shows gauche cis conformation around the alcoholic group.
Benzyl alcohol may be used for the benzylation of toluene.


  • ACS Reagent Grade
  • Presentation: Clear Liquid
  • Auto Ignition: 436 °C (817 °F) (Lit.)
  • Vapor Density: 3.7 (vs air) (Lit.)
  • Vapor Pressure: 0.03 hPa at 20 °C ; 13.3 mmHg ( 100 °C), 3.75 mmHg ( 77 °C) (Lit.)
  • Refractive Index: n20/D 1.539 (Lit.)

Kaava: C₆H₅CH₂OH
Kiehumispiste: 205,4 °C (1013 hPa) (kirj.)
Tiheys: 1,0442 g/cm³ (22.5 °C) (kirj.)
Leimahduspiste: 101 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004599
CAS-numero: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity Test Passes
Color (APHA) ≤20
Residue after Ignition ≤0.005%
Acetophenone ≤0.02%
Benzaldehyde ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR