Selluloosa 97.0-102.0%, microcrystalline powder

Toimittaja: MP Biomedicals

0219149980 0219149983 0219149991
ICNA0219149980EA 37.7 EUR
ICNA0219149980 ICNA0219149983 ICNA0219149991
Selluloosa 97.0-102.0%, microcrystalline powder
Selluloosa

Cellulose is an organic compound, a polysaccharide consisting of a linear chain of several hundred to over ten thousand β(1→4) linked D-glucose units.


  • Water soluble substances 12,0 mg/5 g


Cellulose is used as bulking agent, opacifier, anti-caking agent, extrusion aid and stabiliser for foams and emulsions. High purity cellulose powders are used for partition chromatography.


Soluble in ether, insoluble in water (Water soluble substances <12,0 mg/5 g)

Kaava: (C₁₂H₂₂O₁₁)n
Kiehumispiste: Bulk Density: 0,6 g/ml (25 °C) (Lit,)
Tiheys: 0,6 g/ml (25 °C) (kirj.)
MDL numero: MFCD00081512
CAS-numero: 9004-34-6

Order Now

Testitulosten erittely

Identity Test Passes Test
Purity 97,0 - 102,0%
Moisture content <5,0%
Particle Size +60 mesh (wt%) <8%
Particle Size +200 mesh (wt%) >45%
pH 5 - 7
Residue on ignition <0,05%
Water Soluble Substances <12,0 mg/5 g
Heavy Metals 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR