Kaliumkloridi 99.0-100.5%, crystalline powder ACS

Toimittaja: MP Biomedicals

02191427.1 02191427.5 0219142701 0219142705
ICNA02191427.1EA 36.4 EUR
ICNA02191427.1 ICNA02191427.5 ICNA0219142701 ICNA0219142705
Kaliumkloridi 99.0-100.5%, crystalline powder ACS
Kaliumkloridi

Soluble in water (1 g/3 ml - clear, colorless solution), boiling water (1 g/1,8 ml), glycerol (1 g/14 ml), or ethanol (1 g/250 ml); insoluble in ether or acetone; hydrochloric acid, sodium or magnesium chlorides diminish its solubility in water.

Kaava: KCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011360
CAS-numero: 7447-40-7

Order Now

Testitulosten erittely

Assay 99,0 - 100,5%
pH 5,4 - 8,6 (5% aq soln)
Insoluble Matter ≤0,005%
Iodide ≤0,002%
Bromide ≤0,01%
Chlorate and Nitrate ≤0,003%
Phosphate ≤5 ppm
Sulphate ≤0,001%
Heavy Metals (Pb) 5 ppm;
Iron ≤3 ppm
Calcium ≤0,002%
Magnesium ≤0,001%
Sodium ≤0,005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR