Kaliumbromidi ≥99.0%, white crystalline powder ACS

Toimittaja: MP Biomedicals
Varoitus

0219142680 0219142690 0219142691
ICNA0219142680EA 23.3 EUR
ICNA0219142680 ICNA0219142690 ICNA0219142691
Kaliumbromidi ≥99.0%, white crystalline powder ACS
Kaliumbromidi
Kaava: KBr
Sulamispiste: ~734 °C (kirj.)
MDL numero: MFCD00011358
CAS-numero: 7758-02-3

Order Now

Testitulosten erittely

Identity Test Passes
Assay ≥99,0%
pH (5% aq soln) 5,0 - 8,8
Insoluble Matter <0,005%
Bromate (BrO3) <0,001%,
Iodate (IO3) <0,001%
Chloride (cl) <0,2%
Iodide (I) <0,001%
Sulfate (SO4) <0,005%
Barium (Ba) <0,002%
Heavy Metals (as Pb) <5 ppm
Iron (Fe) <5 ppm
Calcium (Ca) <0,002%
Magnesium (Mg) <0,001%,
Sodium (Na) <0,02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR