Clomiphene citrate salt, off-white powder

Toimittaja: MP Biomedicals

0219017401 0219017425 0219017405 0219017410
ICNA0219017401EA 29.4 EUR
ICNA0219017401 ICNA0219017425 ICNA0219017405 ICNA0219017410
Clomiphene citrate salt, off-white powder
Clomiphene citrate salt

Clomiphene Citrate Salt is a mixture of cis- and trans- isomers. Clomiphene citrate is a selective estrogen receptor modulator.


Freely soluble in methanol; slightly soluble in water and chloroform; sparingly soluble in ethanol (96%); practically insoluble in ether.

Kaava: C₃₂H₃₆ClNO₈
Storage Temperature: Jääkaappi
MDL numero: MFCD00058322
CAS-numero: 50-41-9

Order Now

Testitulosten erittely

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR