Probucol ≥98%

Toimittaja: MP Biomedicals

0215637105 0215637110 0215637150
ICNA0215637105EA 144 EUR
ICNA0215637105 ICNA0215637110 ICNA0215637150
Probucol ≥98%
Probucol
Kaava: C₃₁H₄₈O₂S₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00079281
CAS-numero: 23288-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR