Benfluorex hydrochloride, white powder

Toimittaja: MP Biomedicals

0215482050 0215482010
ICNA0215482050EA 276 EUR
ICNA0215482050 ICNA0215482010
Benfluorex hydrochloride, white powder
Benfluorex hydrochloride
Kaava: C₁₉H₂₁ClF₃NO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00058493
CAS-numero: 23642-66-2
EINECS: 245-777-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR