Nortriptyline hydrochloride, white powder

Toimittaja: MP Biomedicals
Varoitus

0215355583 0215355591
ICNA0215355583EA 46.5 EUR
ICNA0215355583 ICNA0215355591
Nortriptyline hydrochloride, white powder
Nortriptyline hydrochloride

Nortriptyline Hydrochloride is a second-generation tricyclic antidepressant.


Nortriptyline hydrochloride is a more potent inhibitor of the norepinephrine transporter (Ki = 3.4 nM) than the serotonin transporter (Ki = 161 nM). 5-HT2 serotonin receptor antagonist.


Soluble in water, chloroform; sparingly soluble in methal, ethanol (100 mg/mL); practically insoluble in ether, benzene.


Varoitus: Research Use Only (RUO).

Kaava: C₁₉H₂₁N
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00058024
CAS-numero: 894-71-3

Order Now

Testitulosten erittely

Identity Test Passes
Melting Point 213 - 217 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR