Metyylipunainen, dark red powder ACS

Toimittaja: MP Biomedicals

0215254580 0215254583
ICNA0215254580EA 102 EUR
ICNA0215254580 ICNA0215254583
Metyylipunainen, dark red powder ACS
Metyylipunainen

Methyl red is used in the Methyl Red (MR) Test, used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.


  • Visual transition interval from pH 4,2 (pink) to 6,2 (yellow)


Methyl red is used in the methyl red (MR) test, used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.


Soluble in ethanol.


Varoitus: Research Use Only (RUO).

Kaava: C₁₅H₁₅N₃O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002425
CAS-numero: 493-52-7

Order Now

Testitulosten erittely

Melting Point 179 - 182 °C
Clarity of Alcohol Solution Passes Test
Visual Transition Interval From pH 4.2 (pink) to 6.2 (yellow)

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR