2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder

Toimittaja: MP Biomedicals
Vaara

0215079410 0215079425 0215079480
ICNA0215079410EA 73 EUR
ICNA0215079410 ICNA0215079425 ICNA0215079480
2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
2'-Deoxyadenosine-5'-diphosphate trisodium salt

2'-Deoxyadenosine 5'-diphosphate (dADP) may be used for the synthesis of deoxyadenylate oligonucleotides with enzymes such as polynucleotide phosphorylase from Escherichia coli.


dADP is used as an inhibitor of bacterial poly(A) polymerase. DeoxyADP may be use as an alternative substrate or inhibitor for studies of ATPase and DNA and RNA polymerase specificity.


Varoitus: Research Use Only (RUO).

Kaava: C₁₀H₁₃N₅Na₂O₉P₂
Storage Temperature: Pakastin
MDL numero: MFCD00083610
CAS-numero: 72003-83-9

Order Now

Testitulosten erittely

Identity Test Passes
Purity ≥97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR