Kanelialdehydi ≥99%, liquid

Toimittaja: MP Biomedicals
Varoitus

0215069080 0215069090
ICNA0215069080EA 88.4 EUR
ICNA0215069080 ICNA0215069090
Kanelialdehydi ≥99%, liquid
Kanelialdehydi

FEMA Number 2286, Flavis number 5.014 , Council of Europe no. 102c, Organoleptic: cinnamon; clove; spicy, allergen: no known allergens.


  • Presentation: Liquid
  • Vapor Pressure: 2.25 hPa at 20 °C (Lit.)
  • Refractive Index: n20/D 1.621(Lit.)
  • Solubility: Misc with alcohol, ether, chloroform, oils.
  • Storage temperature: +4°C, store under nitrogen

Kaava: C₉H₈O
Kiehumispiste: 246 °C (1014 hPa) (kirj.)
Sulamispiste: –7,5 °C (kirj.)
Tiheys: 1,0497 g/cm³ (20 °C) (kirj.)
Storage Temperature: Jääkaappi
MDL numero: MFCD00007000
CAS-numero: 104-55-2

Order Now

Testitulosten erittely

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR