Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid

Toimittaja: MP Biomedicals
Vaara

0210326325 0210326380
ICNA0210326325EA 95.8 EUR
ICNA0210326325 ICNA0210326380
Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid
Vitamin A palmitate

  • Presentation : Clear Yellow Liquid


Storage: under nitrogen.

Kaava: C₃₆H₆₀O₂
Sulamispiste: 28,..29 °C (kirj.)
Leimahduspiste: 194 °C (381,2 °F) (kirj.)
Storage Temperature: Jääkaappi
MDL numero: MFCD00019414
CAS-numero: 79-81-2

Order Now

Testitulosten erittely

Identity Test Passes
Activity >1,000,000 IU/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR