Tina(II)klorididihydraatii, white crystalline powder

Toimittaja: MP Biomedicals
Vaara

0210294980 0210294990 0210294991
ICNA0210294980EA 79.5 EUR
ICNA0210294980 ICNA0210294990 ICNA0210294991
Tina(II)klorididihydraatii, white crystalline powder
Tina(II)klorididihydraatii

Soluble in hydrochloride.

Kaava: SnCl₂·2H₂O
MDL numero: MFCD00149863
CAS-numero: 10025-69-1
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR