Rubidiumkloridi ≥99%, white powder, Optical Grade

Toimittaja: MP Biomedicals

0210282510 0210282505 0210282550 0210282580
ICNA0210282510EA 107 EUR
ICNA0210282510 ICNA0210282505 ICNA0210282550 ICNA0210282580
Rubidiumkloridi ≥99%, white powder, Optical Grade
Rubidiumkloridi

Rubidium chloride is used as a catalyst and additive in gasoline.


In materials science, RbCl has been used to prepare molecular nanowires as potential precursors of nanoscale devices. In chromatography, RbCl and other univalent salts have been studied for their influence on the capillary electrophoretic separation of amino acids labeled with 3-(4-carboxybenzoyl)-quinoline-2-carboxaldehyde. RbCl is utilised in the transfection of bacteria. RbCl has been used to investigate the gating and permeability of ion channels produced by botulinum neurotoxin types A and E in membranes from cultured PC12 cells.

Kaava: RbCl
Sulamispiste: 715 °C (kirj.)
MDL numero: MFCD00011187
CAS-numero: 7791-11-9

Order Now

Testitulosten erittely

Identity Test Passes
Purity ≥99%
OD280 (40% aq soln) ≤0,2
Solubility (40% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR