Pyridoksiinihydrokloridi ≥98%, white powder

Toimittaja: MP Biomedicals

0210277710 0210277750 0210277780
ICNA0210277710EA 39.7 EUR
ICNA0210277710 ICNA0210277750 ICNA0210277780
Pyridoksiinihydrokloridi ≥98%, white powder
Pyridoksiinihydrokloridi

Pyridoxine Hydrochloride is one of the vitamins of the B6 complex; present in many foodstuffs (yeast, liver and cereals).


Pyridoxine hydrochloride has a role in transporting protein Bsu1, and contributes to thiamine uptake.


Soluble in water (1 g/4,5 ml - clear, colourless to yellow solution), ethanol (1 g/90 ml), propylene glycol; sparingly soluble in acetone; insoluble in ether or chloroform.

Kaava: C₈H₁₂ClNO₃
Sulamispiste: 202…214 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012807
CAS-numero: 58-56-0
EINECS: 200-386-2
Merck Index: 12,08166

Order Now

Testitulosten erittely

Identity Test Passes
Purity ≥98,0%
Melting Point 202 - 214 °C
FTIR Conforms to Standard
pH (10% aq soln) 2,0 - 4,0
Loss on Drying <0,5%
Residue on Ignition <0,1%
Heavy Metals <0,003%
Chloride content 16,9 - 17,6 (dried basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR