Pyridoxal hydrochloride, off-white powder

Toimittaja: MP Biomedicals

0210276725 0210276780 0210276701 0210276705
ICNA0210276725EA 250 EUR
ICNA0210276725 ICNA0210276780 ICNA0210276701 ICNA0210276705
Pyridoxal hydrochloride, off-white powder
Pyridoxal hydrochloride

Pyridoxal hydrochloride is used for labeling amino acids and for their detection in the picomole range.


Finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.


Pyridoxal Hydrochloride is used for labeling amino acids and for their detection in the picomole range.


Soluble in water (0,1% aq soln, 15 min., clear, colourless solution) and in 95% alcohol.


Varoitus: Store Desiccated. Protect from light. Gradually darkens on exposure to air and sunlight

Kaava: C₈H₁₀ClNO₃
Sulamispiste: 170…180 °C
Storage Temperature: Pakastin
MDL numero: MFCD00012809
CAS-numero: 65-22-5
EINECS: 200-602-5

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
Solubility Test (0.1% aq soln, 15 min.) Clear, Colorless Solution C
pH (0,1% aq soln) 2,7 - 3,5
UV/VIS l max (water) 253-5, 290-1, 316-8 nm
Melting Point 170 - 180°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR