Naphthol AS-D chloroacetate ≥98% (by TLC), jauhe

Toimittaja: MP Biomedicals

0210242401 02102424.1 02102424.5
ICNA0210242401EA 320 EUR
ICNA0210242401 ICNA02102424.1 ICNA02102424.5
Naphthol AS-D chloroacetate ≥98% (by TLC), jauhe
Naphthol AS-D chloroacetate

Naphthol AS-D chloroacetate is a substrate for histochemical localization of esterases.


  • Presentation: White Powder


Soluble in ethyl acetate (5 mg/mL - clear, slightly yellow solution); methanol (1 mg/mL-clear, colorless solution) and acetone.

Kaava: C₂₀H₁₆ClNO₃
Sulamispiste: 161…171 °C
Storage Temperature: Pakastin
CAS-numero: 35245-26-2
EINECS: 252-463-5

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
Melting Point 161 - 171°C
UV/Vis: l max (methanol) 232 ± 5 nm
Free Naphthol ≤100 ppm
Purity (TLC) >98%
Solubility (5% in acetone) Clear, Colorless Solution

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR