MOPS, white powder

Toimittaja: MP Biomedicals

0210237010 0210237080 0210237083 0210237091
ICNA0210237010EA 36.5 EUR
ICNA0210237010 ICNA0210237080 ICNA0210237083 ICNA0210237091
MOPS, white powder
MOPS

MOPS is a zwitterionic amino acid which acts as one of the "Good" buffers.


MOPS is used as a biological buffer.


Concentrations higher than 20 mM should not be used with mammalian cell cultures.


Soluble in water (to approximately 33% w/w - clear, colorless solution)


Varoitus: Research Use Only (RUO).

Kaava: C₇H₁₅NO₄S
Sulamispiste: 277 °C (kirj.)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006183
CAS-numero: 1132-61-2

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
UV/Vis: (1M aq soln) OD250 nm ≤0.04
UV/Vis: (1M aq soln) OD280 nm ≤0.04
pH (10% aq soln) 3.2 - 4.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR