3-(Indol-3-yyli)propaanihappo ~99%, jauhe

Toimittaja: MP Biomedicals

0210204810 0210204805 0210204825
ICNA0210204810EA 53.5 EUR
ICNA0210204810 ICNA0210204805 ICNA0210204825
3-(Indol-3-yyli)propaanihappo ~99%, jauhe
3-(Indol-3-yyli)propaanihappo

Indole-3-Propionic Acid is used as reactant for preparation of fluorescent analogues of strigolactones,anti-tumor agents,melanocortin receptors ligands, Immunosuppressive agents, Iinhibitors of hepatitis C virus, Histamine H4 receptor agonists, NR2B/NMDA receptor antagonists, CB1 Antagonist for the treatment of obesity, Antibacterial agents, Inhibitor of TGF-β receptor binding.

Kaava: C₁₁H₁₁NO₂
Sulamispiste: 129…132 °C
Tiheys: 1,356 g/cm³ (20 °C)
MDL numero: MFCD00005660
CAS-numero: 830-96-6
EINECS: 212-600-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR