Imidatsoli ≥99%, white crystalline powder

Toimittaja: MP Biomedicals
Vaara

02102033.1 02102033.5 0210203301 0210203305
ICNA02102033.1EA 59.4 EUR
ICNA02102033.1 ICNA02102033.5 ICNA0210203301 ICNA0210203305
Imidatsoli ≥99%, white crystalline powder
Imidatsoli

Imidazole can be used for the elution of histidine containing proteins from divalent cation resins and can also be used in reverse staining of SDS-PAGE gels for detection of proteins .


  • Excellent for buffer use in pH range 6.2 - 7.8


Imidazole is used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column. Synthetic imidazoles are present in many fungicides and antifungal, antiprotozoal, and antihypertensive medications.


Freely soluble in water, alcohol, ehter, chloroform, pyridine; slightly soluble in benzene; very sparingly soluble in petroleum ether


Varoitus: Research Use Only (RUO).

Kaava: C₃H₄N₂
Kiehumispiste: 268 °C (kirj.)
Sulamispiste: 86…94 °C
Tiheys: 500…600 kg/m³ (bulk density)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005183
CAS-numero: 288-32-4
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity ≥99%
FTIR Conforms to Standard
Melting Point 86 - 94 °C
pH (1% aq.) ≥7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR