L(+)-glutamiinihappo ≥99%, white powder

Toimittaja: MP Biomedicals

02101793.1 0210179301 0210179305
ICNA02101793.1EA 29.2 EUR
ICNA02101793.1 ICNA0210179301 ICNA0210179305
L(+)-glutamiinihappo ≥99%, white powder
L(+)-glutamiinihappo

Glutamate is a key compound in cellular metabolism. It is the most abundant excitatory neurotransmitter in the vertebrate nervous system. It also serves as the precursor for the synthesis of the inhibitory GABA in GABA-ergic neurons. This reaction is catalyzed by glutamate decarboxylase (GAD), which is most abundant in the cerebellum and pancreas.


It is often used as a food additive and flavour enhancer in the form of its salt, known as monosodium glutamate (MSG). A glutamic acid derivative, poly-γ-benzyl-L-glutamate (PBLG), is often used as an alignment medium to control the scale of the dipolar interactions observed. L-Glutamic acid is used in cell culture as a component of MEM non-essential amino acids solution. L-Glutamic acid has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production. The competitive roles of L-glutamic acid and L-aspartic acid in the growth of Oenococcus oeni NCFB 1707 have been studied.


Clear, Colorless Solution (10% in 2N HCl). Soluble in water (8.64 g/L at room temperature; 140 g/L in boiling water); insoluble in methanol, ethanol, ether, acetone, cold glacial acetic acid and common neutral solvents.

Kaava: HO₂CCH₂CH₂CH(NH₂)CO₂H
Kiehumispiste: 325 °C (1013 hPa)
Sulamispiste: 160 °C
Tiheys: 1,538 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002634
CAS-numero: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Moisture Content ≤0.15%
Optical Rotation (c=10, 2N HCl ) +25° to +35°
Solubility (10% in 2N HCl) Clear, Colorless Solution
Arsenic ≤1,5 ppm
Heavy Metals ≤20 ppm (as Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR