Histamine diphosphate, crystalline powder

Toimittaja: MP Biomedicals
Vaara

0210034301 0210034305 0210034325
ICNA0210034301EA 41 EUR
ICNA0210034301 ICNA0210034305 ICNA0210034325
Histamine diphosphate, crystalline powder
Histamine diphosphate

Histamine Diphosphate Monohydrate is potent vasodilater.


Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2â‚‚ activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.


Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilises Ca2+; H22‚ activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.


Soluble in water (50 mg/mL - clear, colorless to almost colorless solution).


Varoitus: Research Use Only (RUO).

Kaava: C₅H₉N₃·2H₃PO₄
Storage Temperature: Pakastin
MDL numero: MFCD00064316
CAS-numero: 51-74-1

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
Loss on Drying Report result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR