Nikotiiiniamidiadeniinidinukleotidi (hapettunut) ≥93%

Toimittaja: MP Biomedicals

0210031905 0210031901 0210031925
ICNA0210031905EA 211 EUR
ICNA0210031905 ICNA0210031901 ICNA0210031925
Nikotiiiniamidiadeniinidinukleotidi (hapettunut) ≥93%
Nikotiiiniamidiadeniinidinukleotidi (hapettunut)

This grade of NAD is intended for applications where maximum reagent purity is not critical. Well suited for use on most automatic analysers.


  • White powder, soluble in water (200 mg/ml)
  • pH: 2 - 4 (1% aqueous solution)


Solutions should be aliquoted and stored at –20 °C for up to one week.

Kaava: C₂₁H₂₇N₇O₁₄P₂
MDL numero: MFCD00036253
CAS-numero: 53-84-9

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR