Alumiinioksidi, Ecochrom™ Alumina B Activity: Super I

Toimittaja: MP Biomedicals

0204574
ICNA0204574EA 338 EUR
ICNA0204574
Alumiinioksidi, Ecochrom™ Alumina B Activity: Super I
Alumiinioksidi

Specific Surface Area: 200 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 10


pH: 10

Kaava: Al₂O₃
Tiheys: 3,94 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003424
CAS-numero: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR