Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Toimittaja: Thermo Fisher Scientific
CHSC9-10 CHSC9-10F CHSC9-10T CHSC9-20 CHSC9-20F CHSC9-20T CHSC9-30 CHSC9-30F CHSC9-30T CHSC9-35UB CHSC9-35UBF CHSC9-35UBS CHSC9-35UBSF CHSC9-35UBST CHSC9-35UBT CHSC9-40 CHSC9-40F CHSC9-40T
HYPECHSC9-10EA 18 EUR
HYPECHSC9-10 HYPECHSC9-10F HYPECHSC9-10T HYPECHSC9-20 HYPECHSC9-20F HYPECHSC9-20T HYPECHSC9-30 HYPECHSC9-30F HYPECHSC9-30T HYPECHSC9-35UB HYPECHSC9-35UBF HYPECHSC9-35UBS HYPECHSC9-35UBSF HYPECHSC9-35UBST HYPECHSC9-35UBT HYPECHSC9-40 HYPECHSC9-40F HYPECHSC9-40T
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Tulpat ja korkit Kromatografian näytekuppien tulpat
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including our SureSTART 2 ml Glass Screw Vials and 2 ml polypropylene screw Microvials for <2 ml samples (Level 1). Select level 1 caps for chromatography approved, cost optimised products that are suitable for everyday analyses and compatible with all instrument types.
Choose from polypropylene caps with silicone/PTFE or rubber/TEF septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate advanced vial closure systemMore Product Information