HPLC column filters, direct-connection, Javelin™
Toimittaja: Thermo Fisher Scientific
88200 88400 88700
HYPE88200EA 250 EUR
HYPE88200 HYPE88400 HYPE88700
HPLC column filters, direct-connection, Javelin™
Suodattimet Esikolonnit ja fritit
Protect HPLC systems with one-piece Javelin™ direct-connection column filters.
  • Replace entire disposable filter unit for easy changes
  • Recommended for use as dedicated filters for a column rather than the HPLC system
  • CPI tip attaches directly to HPLC column inlet without tubing or wrenches