Glass snap top microvials, 0,3 ml for <2 ml samples, level 3 high performance applications, SureSTART™
Toimittaja: Thermo Fisher Scientific
6PRV11-03FIVP 6PRV11-03FIVPF 6PRV11-03FIVPT
HYPE6PRV11-03FIVPEA 101 EUR
HYPE6PRV11-03FIVP HYPE6PRV11-03FIVPF HYPE6PRV11-03FIVPT
Glass snap top microvials, 0,3 ml for <2 ml samples, level 3 high performance applications, SureSTART™
Näytevialit ja pienet näyteastiat
Choose Thermo Scientific™ SureSTART™ glass snap top microvials, performance level 3, to maximise the injection volume when analysing <2 ml samples.
With outer dimensions of 12×32 mm, this clear or amber glass microvial has a conical-shaped, fixed inner insert to enable a usable volume of 0.25 ml and a residual volume of 4 µl. This is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are specification certified to ensure physical dimensions and autosamplerMore Product Information