Snap caps, 11 mm, level 3 high performance applications, SureSTART™
Toimittaja: Thermo Fisher Scientific
6PRC11ST1 6PRC11ST101R 6PRC11ST101RF 6PRC11ST101RT 6PRC11ST1F 6PRC11ST1R 6PRC11ST1RF 6PRC11ST1RT 6PRC11ST1T 6PRC11STS1 6PRC11STS1F 6PRC11STS1R 6PRC11STS1RF 6PRC11STS1RT 6PRC11STS1T 6PRC11STS1X 6PRC11STS1XF 6PRC11STS1XT 6PRC11ST101 6PRC11ST101F 6PRC11ST101T
HYPE6PRC11ST1EA 0 EUR
HYPE6PRC11ST1 HYPE6PRC11ST101R HYPE6PRC11ST101RF HYPE6PRC11ST101RT HYPE6PRC11ST1F HYPE6PRC11ST1R HYPE6PRC11ST1RF HYPE6PRC11ST1RT HYPE6PRC11ST1T HYPE6PRC11STS1 HYPE6PRC11STS1F HYPE6PRC11STS1R HYPE6PRC11STS1RF HYPE6PRC11STS1RT HYPE6PRC11STS1T HYPE6PRC11STS1X HYPE6PRC11STS1XF HYPE6PRC11STS1XT HYPE6PRC11ST101 HYPE6PRC11ST101F HYPE6PRC11ST101T
Snap caps, 11 mm, level 3 high performance applications, SureSTART™
Tulpat ja korkit Kromatografian näytekuppien tulpat
Use Thermo Scientific™ SureSTART™ 11 mm Snap caps with snap vials that have an 11 mm opening, including our SureSTART 2 ml glass snap Vials, 1.5 ml total recovery glass snap vials, high recovery glass snap vials, and glass snap micro vials for <2 ml samples (level 3).
Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Choose from soft polyethylene caps with double snap functionality that guarantees a tight seal.
These snap caps include all-purpose silicone/PTFE septa of variousMore Product Information