SPE plates, 96-well, HyperSep™ Phenyl
Toimittaja: Thermo Fisher Scientific
60300-688
HYPE60300-688EA 537 EUR
HYPE60300-688
SPE plates, 96-well, HyperSep™ Phenyl
Kuoppalevyt Kiinteäfaasiuuttolevyt
In Thermo Scientific™ HyperSep™ Phenyl Plates, the electron density of the aromatic ring provides different selectivity than other nonpolar sorbents for retention of basic compounds.
The cartridges are commonly used for nonpolar interactions, such as:
- Extraction of aromatic compounds
- Extraction of basic analytes