Terry tool tubing cutters
Toimittaja: Thermo Fisher Scientific
60182-510
HYPE60182-510EA 473 EUR
HYPE60182-510
Terry tool tubing cutters
Leikkurit
Tubing cutter cuts ¹/₈" stainless steel tubing.
Produce clean, 90° cuts of ¹/₈" stainless steel tubing.