SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
Toimittaja: Thermo Fisher Scientific
60180-P330 60180-P332 60180-P334 60180-P336 60180-P338 60180-P341 60180-P343 60180-P350 60180-P330B 60180-P332B 60180-P334B 60180-P336B 60180-P341B 60180-P350B 60180-P338B 60180-P343B
HYPE60180-P330EA 574 EUR
HYPE60180-P330 HYPE60180-P332 HYPE60180-P334 HYPE60180-P336 HYPE60180-P338 HYPE60180-P341 HYPE60180-P343 HYPE60180-P350 HYPE60180-P330B HYPE60180-P332B HYPE60180-P334B HYPE60180-P336B HYPE60180-P341B HYPE60180-P350B HYPE60180-P338B HYPE60180-P343B
SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
Kuoppalevyt Solid Supported Liquid/Liquid Extraction Plates (SLE)
SureSTART™ Level 3 WebSeal™ Plate 96-well round U, V, and flat-bottom microtiter plates are designed for regulated labs that require high performing products. Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
  • Specification certified
  • Suitable for HPLC/UHPLC applications
  • Performance Level 3 products include unique WebSeal Plate+ glass-coated polypropylene plates and plates with glass inserts
  • Suitable for QA/QC applications using core detectors such as UV, DAD/