Positive pressure manifolds, HyperSep™
Toimittaja: Thermo Fisher Scientific
60104-236 60104-274
HYPE60104-236EA 10100 EUR
HYPE60104-236 HYPE60104-274
Positive pressure manifolds, HyperSep™
Manifoldit
HyperSep™ positive pressure manifold can hold 48 SPE cartridges of various sizes 1, 3, 6, and 10/15 ml. Samples and solvent added to the SPE cartridges are extracted under positive pressure for an even flow at each position. Robust way to prepare multiple samples for subsequent analysis by chromatography.
  • Extracts are collected into 13 or 16 mm tubes