Cell culture media, liquid, DMEM:F12 (1:1), HyClone™
Toimittaja: HyClone products (Cytiva)
SH30261.01 SH30023.01 SH30023.02 SH30023.FS SH30126.FS SH30271.01 SH30271.FS SH30272.02
SH30261.01EA 25.9 EUR
SH30261.01 HYCLSH30023.01 HYCLSH30023.02 HYCLSH30023.FS HYCLSH30126.FS HYCLSH30271.01 HYCLSH30271.FS HYCLSH30272.02
Cell culture media, liquid, DMEM:F12 (1:1), HyClone™
Soluviljelyyn liuokset
HyClone Classical Liquid Media: DMEM F12 is manufactured using ISO 9001:2000-certified processes.
  • Contains 17,5 mM glucose
  • 0,1 µm sterile filtered
  • Manufactured using ISO 9001 certified processes
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as raw material components must pass strict Quality Control testing to confirm the appropriate level of quality.
Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection. We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.