Wire baskets, rectangular, with lid
Toimittaja: HUPFER METALLWERKE
7501132 7501135 7501133 7501204 7501161 7501162 7501282
HUPF7501132EA 273 EUR
HUPF7501132 HUPF7501135 HUPF7501133 HUPF7501204 HUPF7501161 HUPF7501162 HUPF7501282
Wire baskets, rectangular, with lid
Korit Korit, lanka
These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.
  • With lid
  • With base frame
  • Mesh size: 1,6×1,6 mm
Colour: Silver.