Container, with screw cap
Toimittaja: hünersdorff GmbH
915200 915300 915600
HUEN915200EA 0 EUR
HUEN915200 HUEN915300 HUEN915600
Container, with screw cap
Näyteastiat, muovia Yleiskäyttöinenastiat
Containers with screw cap round, for varied applications, made of food-safe natural polypropylene.
  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable