Bath fluids
Toimittaja: HUBER
6158 6673 6164 6162 6259 6258 6479 6161 6155 6156 6157 6163 6165 6166 6167 6168 6170 6171 6275 6276 6672 9685 10656
HUBE6158EA 886 EUR
HUBE6158 HUBE6673 HUBE6164 HUBE6162 HUBE6259 HUBE6258 HUBE6479 HUBE6161 HUBE6155 HUBE6156 HUBE6157 HUBE6163 HUBE6165 HUBE6166 HUBE6167 HUBE6168 HUBE6170 HUBE6171 HUBE6275 HUBE6276 HUBE6672 HUBE9685 HUBE10656
Bath fluids
Haudenesteet ja haudeöljyt
Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.
All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.
Varoitus: * –20...+235 °C for closed system applications
Use biocides safely. Always read the label and product information before use