Clinisafe® 5th Generation Recycled Containers
Toimittaja: Hospidex
HOS30LPBRLWPH5 HOS50LPBRLWPH5 HOS60LPBRLWPH5 HOS30LPBRCW4 HOS50LPBRCW4 HOS60LPBRCW4
HUARHOS30LPBRLWPH5EA 0 EUR
HUARHOS30LPBRLWPH5 HUARHOS50LPBRLWPH5 HUARHOS60LPBRLWPH5 HUARHOS30LPBRCW4 HUARHOS50LPBRCW4 HUARHOS60LPBRCW4
Clinisafe® 5th Generation Recycled Containers
Jäteastiat ja -säiliöt Sharps Containers
Made from 30% recycled plastic. Fully opening lid, with original positioning of hinge.
  • Permanent closure indicator: visual and very easy to check
  • Specific permanent closure with independent safety tabs: can be activated in any order
  • Lid/base joint seal system always suitably adjusted
  • External dimensions suitable for placement on a 1200×800 mm pallet enabling optimal collection for transport to the waste treatment centre
  • Tested for autoclave resistance (open lid, before use)