Sigma-Aldrich® Bouin's solution
Toimittaja: SIGMA ALDRICH MICROSCOPY
HT101128-4L HT10132-1L
HT101128-4LEA 409 EUR
HT101128-4L HT10132-1L
Sigma-Aldrich® Bouin's solution
Värit Microscopy and staining reagents
Bouin's solution is an excellent fixative for preserving soft and delicate structures. It is prepared with saturated picric acid, formaldehyde and acetic acid.
  • Acetic acid 5%
  • Formaldehyde 9%
  • Picric acid 0,9%
Used as a mordant in various trichrome procedures.