Magneettisekoittaja, Variomag Mobil 600
Toimittaja: THERMO H + P LABORTECHNIK
50119110
HPLT50119110EA 4070 EUR
HPLT50119110
Magneettisekoittaja, Variomag Mobil 600
Sekoittajat Magneettisekoittajat
Magnetic stirrer with external control, units are ideal for stirring large volumes and integration in analytical instruments and systems.
  • Rugged construction with stainless steel housing, units can be cleaned under running water
  • Safety features include overload protection, quick-stop function which stops drive within three seconds and adjustable automatic start up for safe stirrer acceleration
  • Can be used at −10 to +56 °C (100% relative humidity)
  • Telemodul 80 M control unit allows speed to be adjusted between 100 to 1000 min⁻¹ and ensures constant speed