Aflatoxin Mix 4-solution 2 µg/mL B1+G1 and 0.5 µg/mL B2+G2 in acetonitrile, Analytical standard, reference material, Fluka™
Toimittaja: Honeywell Chemicals
R1961-2ML
HONCR1961-2MLEA 396.94 EUR
HONCR1961-2ML
Aflatoxin Mix 4-solution 2 µg/mL B1+G1 and 0.5 µg/mL B2+G2 in acetonitrile, Analytical standard, reference material, Fluka™
Standardiaineet ja vertailumateriaalit Organic Standards
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery