, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Toimittaja: Honeywell Chemicals
Vaara

LC441-2.5
HONCLC441-2.5EA 1350 EUR
HONCLC441-2.5
, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Asetonitriili kanssa 0,1% (v/v) Muurahaishappo

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Kiehumispiste: 82 °C (1,013 hPa)
Sulamispiste: –46 °C
Tiheys: 0,7767 g/cm³ (25 °C)
Leimahduspiste: 8 °C
Storage Temperature: Huoneenlämpö
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR