Thallium AAS Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E4078-100ML E4078-500ML
HONCE4078-100MLEA 39.13 EUR
HONCE4078-100ML HONCE4078-500ML
Thallium AAS Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Thallium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).