Manganese AAS standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E4044-100ML E4044-500ML E4045-100ML E4045-500ML
HONCE4044-100MLEA 34.01 EUR
HONCE4044-100ML HONCE4044-500ML HONCE4045-100ML HONCE4045-500ML
Manganese AAS standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Manganese Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).