Barium AAS Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E4007-100ML E4007-500ML E4008-100ML E4008-500ML E4106-100ML
HONCE4007-100MLEA 34.58 EUR
HONCE4007-100ML HONCE4007-500ML HONCE4008-100ML HONCE4008-500ML HONCE4106-100ML
Barium AAS Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Barium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).