Monoethanolamine standard for ion chromatography, (CRM), Fluka™
Toimittaja: Honeywell Chemicals
E3029-100ML
HONCE3029-100MLEA 84.34 EUR
HONCE3029-100ML
Monoethanolamine standard for ion chromatography, (CRM), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Amine Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).